siRNA Details
| Gene Symbol: Ndufa1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019443.2 | NP_062316.1 | 3 | 384 | ||
| GenBank mRNA | AK007840.1 | - | 3 | 291 | ||
| AK131584.1 | - | 583 | ||||
| BC024364.1 | - | 3 | 293 | 5031206 | ||
| CF383819.1 | - | 2 | 168 | |||
| Y07708.1 | - | 3 | 403 | |||

Ndufa1
Genomic Map