siRNA Details
| Gene Symbol: Nfat5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001286260.1 | NP_001273189.1 | 12 | 3760 | ||
| NM_018823.2 | NP_061293.2 | 13 | 3814 | |||
| NM_133957.3 | NP_598718.2 | 14 | 3876 | |||
| GenBank mRNA | AF162853.1 | - | 12 | 3671 | ||
| AF200687.1 | - | 13 | 3449 | |||
| AF369980.1 | - | 14 | 3515 | |||
| AF453571.2 | - | 14 | 3484 | |||
| AK122381.1 | - | 4 | 1901 | |||

Nfat5
Genomic Map