siRNA Details
| Gene Symbol: Il1f6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019450.3 | NP_062323.1 | 4 | 393 | ||
| GenBank mRNA | AF200493.1 | - | 3 | 232 | ||
| AF206697.1 | - | 4 | 307 | |||
| AK004061.1 | - | 4 | 393 | |||
| AK224931.1 | - | 4 | 327 | |||
| AK224932.1 | - | 4 | 340 | |||
| BC117102.1 | - | 3 | 266 | 40126421 | ||
| BC117104.1 | - | 3 | 266 | 40126423 | ||

Il1f6
Genomic Map