siRNA Details
| Gene Symbol: Cma2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024714.2 | NP_001019885.1 | 2 | 168 | ||
| GenBank mRNA | AF361939.1 | - | 2 | 168 | ||
| BC139236.1 | - | 2 | 163 | 8862258 | ||
| BC139237.1 | - | 2 | 163 | 8862259 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cma2| Gene Symbol: Cma2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024714.2 | NP_001019885.1 | 2 | 168 | ||
| GenBank mRNA | AF361939.1 | - | 2 | 168 | ||
| BC139236.1 | - | 2 | 163 | 8862258 | ||
| BC139237.1 | - | 2 | 163 | 8862259 | ||