siRNA Details
| Gene Symbol: B020004J07Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033790.3 | NP_001028962.1 | 3 | 861 | ||
| GenBank mRNA | AK139557.1 | - | 3 | 862 | ||
| BC147376.1 | - | 2 | 757 | 9056473 | ||
| BC147377.1 | - | 2 | 757 | 9056471 | ||

B020004J07Rik
Genomic Map