siRNA Details
| Gene Symbol: BMP8B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001720.3 | NP_001711.2 | 7 | 2489 | ||
| XM_011542024.2 | XP_011540326.2 | 8 | 2282 | |||
| GenBank mRNA | AB075489.1 | - | 1 | 529 | ||
| AY303955.1 | - | 7 | 2489 | |||
| BC023526.2 | - | 7 | 2088 | |||

BMP8B
Genomic Map