siRNA Details
| Gene Symbol: TMEM147 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001242597.1 | NP_001229526.1 | 5 | 691 | ||
| NM_001242598.1 | NP_001229527.1 | 4 | 398 | |||
| NM_032635.3 | NP_116024.1 | 6 | 620 | |||
| GenBank mRNA | BC001118.2 | - | 6 | 577 | 2989840 | |
| BG697070.1 | - | 5 | 580 | |||
| BM423809.1 | - | 6 | 620 | |||
| BM765457.1 | - | 4 | 208 | |||
| Y18007.1 | - | 6 | 588 | |||

TMEM147
Genomic Map