siRNA Details
| Gene Symbol: Trim43a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001034906.2 | NP_001030078.1 | 2 | 272 | ||
| GenBank mRNA | AK145647.1 | - | 2 | 273 | ||
| AK166678.1 | - | 2 | 271 | |||
| BC141236.1 | - | 2 | 151 | 9055848 | ||
| Gene Symbol: Trim43b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001170884.1 | NP_001164355.1 | 2 | 417 | ||

Trim43a
Genomic Map