siRNA Details
| Gene Symbol: Cts7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019539.3 | NP_062412.1 | 3, 4 | 473 | ||
| GenBank mRNA | AF250837.1 | - | 2, 3 | 260 | ||
| AK017466.1 | - | 3, 4 | 499 | |||
| AY014779.1 | - | 3, 4 | 333 | |||
| BC064740.1 | - | 3, 4 | 473 | 30018572 | ||

Cts7
Genomic Map