siRNA Details
| Gene Symbol: Plac1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019538.4 | NP_062411.1 | 3 | 349 | ||
| GenBank mRNA | AF234653.1 | - | 3 | 350 | ||
| AF250838.1 | - | 3 | 346 | |||
| AK005501.1 | - | 3 | 350 | |||
| AK145876.1 | - | 3 | 315 | |||
| BC051666.1 | - | 3 | 335 | 30017089 | ||

Plac1
Genomic Map