siRNA Details
| Gene Symbol: Stx1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024414.2 | NP_077725.1 | 10 | 1194 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Stx1b| Gene Symbol: Stx1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024414.2 | NP_077725.1 | 10 | 1194 | ||