siRNA Details
| Gene Symbol: 1110004F10Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019772.2 | NP_062746.1 | 5 | 565 | ||
| GenBank mRNA | AB025407.1 | - | 5 | 565 | ||
| AK003415.1 | - | 5 | 606 | |||
| AK144059.1 | - | 5 | 592 | |||
| AK167571.1 | - | 5 | 594 | |||
| AK168206.1 | - | 5 | 599 | |||
| AK169309.1 | - | 5 | 592 | |||
| AK170172.1 | - | 3 | 323 | |||
| BC072575.1 | - | 5 | 601 | 5715413 | ||

1110004F10Rik
Genomic Map