siRNA Details
| Gene Symbol: Tulp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AF105712.1 | - | 7 | 670 | ||
| AK054304.1 | - | 4 | 628 | |||
| AK077200.1 | - | 7 | 679 | |||
| AK077211.1 | - | 4 | 476 | |||
| AK161165.1 | - | 4 | 627 | |||
| BB617192.1 | - | 7 | 679 | |||
| BC089545.1 | - | 4 | 475 | 6770730 | ||

Tulp2
Genomic Map