siRNA Details
Gene Symbol: C1qtnf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001204129.1 | NP_001191058.1 | 4 | 1422 | ||
NM_001204130.1 | NP_001191059.1 | 4 | 1334 | |||
NM_019959.3 | NP_064343.1 | 4 | 1412 | |||
GenBank mRNA | AF192499.1 | - | 4 | 1360 | ||
AK005484.1 | - | 4 | 1381 | |||
BC023468.1 | - | 1 | 1011 | |||
BC037636.1 | - | 3 | 1327 | |||
BC049894.1 | - | 4 | 1363 | 4018549 |
