siRNA Details
| Gene Symbol: Tssc4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001115085.1 | NP_001108557.1 | 3 | 1113 | ||
| NM_020285.2 | NP_064681.1 | 3 | 1143 | |||
| NM_138631.1 | NP_619537.1 | 4 | 644 | |||
| GenBank mRNA | AB038011.1 | - | 3 | 1143 | ||
| AB041597.1 | - | 1 | 1484 | |||
| AJ279796.1 | - | 3 | 1143 | |||
| AJ279797.1 | - | 4 | 644 | |||
| AK088052.1 | - | 3 | 1077 | |||
| BC131974.1 | - | 3 | 456 | |||
| BC131976.1 | - | 3 | 456 | |||
| BC145365.1 | - | 3 | 456 | |||

Tssc4
Genomic Map