siRNA Details
| Gene Symbol: Psors1c2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020576.2 | NP_065601.2 | 2 | 784 | ||
| GenBank mRNA | AF159091.1 | - | 2 | 716 | ||
| AF484421.1 | - | 2 | 743 | |||
| AK137759.1 | - | 2 | 785 | |||
| BC110561.2 | - | 2 | 712 | 40049760 | ||

Psors1c2
Genomic Map