siRNA Details
| Gene Symbol: IFI30 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_006332.4 | NP_006323.2 | 5, 6 | 696 | ||
| GenBank mRNA | AB049659.1 | - | 5, 6 | 684 | ||
| AF097362.1 | - | 5, 6 | 667 | |||
| AK291551.1 | - | 5, 6 | 681 | |||
| AL558891.3 | - | 5, 6 | 696 | |||
| BC021136.1 | - | 5, 6 | 665 | 4863531 | ||
| BC031020.1 | - | 5, 6 | 663 | 4690088 | ||
| DQ884410.1 | - | 5, 6 | 577 | |||

IFI30
Genomic Map