siRNA Details
| Gene Symbol: Isy1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133934.4 | NP_598695.1 | 3, 4 | 188 | ||
| GenBank mRNA | AK017996.1 | - | 3, 4 | 105 | ||
| AK173116.1 | - | 156 | ||||
| BC020103.1 | - | 3, 4 | 152 | 3673446 | ||
| BC037695.1 | - | 3, 4 | 193 | 4009615 | ||

Isy1
Genomic Map