siRNA Details
| Gene Symbol: Crlf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001164735.1 | NP_001158207.1 | 5 | 726 | ||
| NM_016715.4 | NP_057924.3 | 5 | 726 | |||
| GenBank mRNA | AB031333.1 | - | 5 | 674 | ||
| AB039945.1 | - | 5 | 622 | |||
| AF201963.1 | - | 5 | 777 | |||
| AF232936.1 | - | 5 | 690 | |||
| AK149928.1 | - | 5 | 728 | |||
| AK170466.1 | - | 2 | 588 | |||
| BC016580.1 | - | 5 | 804 | 2655857 | ||
| BC023788.1 | - | 5 | 714 | 5346748 | ||

Crlf2
Genomic Map