siRNA Details
| Gene Symbol: Fam181b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021427.2 | NP_067402.2 | 1 | 435 | ||
| GenBank mRNA | AB041569.1 | - | 1 | 395 | ||
| AK043957.1 | - | 1 | 450 | |||
| AK140685.1 | - | 1 | 387 | |||
| AK141493.1 | - | 1 | 33 | |||
| BC043329.1 | - | 1 | 384 | 5369836 | ||

Fam181b
Genomic Map