siRNA Details
| Gene Symbol: Thap11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021513.2 | NP_067488.1 | 1 | 1481 | ||
| GenBank mRNA | AB041579.1 | - | 1 | 1442 | ||
| AK165981.1 | - | 1 | 1442 | |||
| AK166109.1 | - | 1 | 1478 | |||
| AK170457.1 | - | 1 | 1453 | |||
| BC003949.1 | - | 1 | 1034 | |||
| BC138966.1 | - | 1 | 1393 | 8861988 | ||
| BC138987.1 | - | 1 | 1393 | 8862009 | ||

Thap11
Genomic Map