siRNA Details
| Gene Symbol: Vmn2r116 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001104580.1 | NP_001098050.1 | 3 | 569 | ||
| GenBank mRNA | AB540947.1 | - | 3 | 569 | ||
| Gene Symbol: Vmn2r111 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001104573.1 | NP_001098043.1 | 3 | 572 | ||
| GenBank mRNA | AB540944.1 | - | 3 | 572 | ||
| Gene Symbol: Vmn2r112 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001104575.1 | NP_001098045.1 | 3 | 572 | ||
| GenBank mRNA | AB540943.1 | - | 3 | 572 | ||
More Information
Set Membership: |
|

Vmn2r116
Genomic Map