siRNA Details
| Gene Symbol: C130026I21Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037909.3 | NP_001032998.2 | 7 | 1590 | ||
| NM_175219.3 | NP_780428.1 | 8 | 1674 | |||
| GenBank mRNA | AK047983.1 | - | 8 | 1652 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

C130026I21Rik| Gene Symbol: C130026I21Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037909.3 | NP_001032998.2 | 7 | 1590 | ||
| NM_175219.3 | NP_780428.1 | 8 | 1674 | |||
| GenBank mRNA | AK047983.1 | - | 8 | 1652 | ||