siRNA Details
Gene Symbol: Gm5725 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166711.1 | NP_001160183.1 | 1 | 778 |
Gene Symbol: Gm8720 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166756.1 | NP_001160228.1 | 1 | 778 |
Gene Symbol: Vmn1r129 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166725.1 | NP_001160197.1 | 1 | 778 |
