siRNA Details
| Gene Symbol: Psme2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001281472.1 | NP_001268401.1 | 1 | 660 | ||
| GenBank mRNA | D87910.1 | - | 1 | 387 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Psme2b| Gene Symbol: Psme2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001281472.1 | NP_001268401.1 | 1 | 660 | ||
| GenBank mRNA | D87910.1 | - | 1 | 387 | ||