siRNA Details
| Gene Symbol: Speer4e | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001122661.1 | NP_001116133.1 | 5 | 1010 | ||
| GenBank mRNA | BC100449.1 | - | 956 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Speer4e| Gene Symbol: Speer4e | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001122661.1 | NP_001116133.1 | 5 | 1010 | ||
| GenBank mRNA | BC100449.1 | - | 956 | |||