siRNA Details
Gene Symbol: Zfp951 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039231.3 | NP_001034320.3 | 4 | 615 | ||
GenBank mRNA | AK132470.1 | - | 4 | 619 | ||
CD807221.1 | - | 1 | 49 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Zfp951 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039231.3 | NP_001034320.3 | 4 | 615 | ||
GenBank mRNA | AK132470.1 | - | 4 | 619 | ||
CD807221.1 | - | 1 | 49 |