siRNA Details
| Gene Symbol: Cyld | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001128170.2 | NP_001121642.1 | 9 | 1813 | ||
| NM_001128171.2 | NP_001121643.1 | 9 | 1938 | |||
| NM_173369.3 | NP_775545.1 | 8 | 1804 | |||
| GenBank mRNA | AK039054.1 | - | 10 | 1942 | ||
| AK048183.1 | - | 2 | 358 | |||
| AK122389.1 | - | 9 | 1919 | |||
| AK133479.1 | - | 8 | 1768 | |||
| AK147204.1 | - | 9 | 1782 | |||
| AK147426.1 | - | 9 | 1777 | |||
| BC028885.1 | - | 1 | 1459 | |||
| BC042438.1 | - | 8 | 1757 | 3983771 | ||

Cyld
Genomic Map