siRNA Details
| Gene Symbol: Zfp800 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081678.1 | NP_001075147.1 | 5 | 1918 | ||
| GenBank mRNA | AK036691.1 | - | 6 | 2185 | ||
| BC066073.1 | - | 5 | 2330 | |||
| BC119236.1 | - | 5 | 1918 | 8733985 | ||
| BC119262.1 | - | 5 | 1918 | 8734011 | ||
| BC144761.1 | - | 5 | 1918 | 9053309 | ||

Zfp800
Genomic Map