siRNA Details
| Gene Symbol: Zfp808 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039239.2 | NP_001034328.2 | 4 | 1405 | ||
| GenBank mRNA | BC061203.1 | - | 4 | 1405 | 30248220 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zfp808| Gene Symbol: Zfp808 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039239.2 | NP_001034328.2 | 4 | 1405 | ||
| GenBank mRNA | BC061203.1 | - | 4 | 1405 | 30248220 | |