siRNA Details
| Gene Symbol: BMX | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001320866.1 | NP_001307795.1 | 7 | 744 | ||
| NM_001721.6 | NP_001712.1 | 7 | 682 | |||
| NM_203281.2 | NP_975010.1 | 7 | 744 | |||
| XM_017029752.1 | XP_016885241.1 | 7 | 714 | |||
| GenBank mRNA | AF045459.1 | - | 7 | 758 | ||
| AK291824.1 | - | 7 | 714 | |||
| AK313568.1 | - | 7 | 677 | |||
| BC016652.2 | - | 7 | 699 | 3919253 | ||
| BQ226991.1 | - | 7 | 704 | |||
| X83107.1 | - | 7 | 604 | |||

BMX
Genomic Map