siRNA Details
| Gene Symbol: Arl4c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177305.4 | NP_796279.2 | 1 | 864 | ||
| GenBank mRNA | AK080335.1 | - | 1 | 808 | ||
| AK080380.1 | - | 1 | 808 | |||
| AK169890.1 | - | 1 | 301 | |||
| BC049804.1 | - | 549 | ||||
| BC055769.1 | - | 1 | 824 | 6414771 | ||
| BC115952.1 | - | 1 | 44 | |||

Arl4c
Genomic Map