siRNA Details
| Gene Symbol: SLC30A9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_006345.3 | NP_006336.3 | 3 | 434 | ||
| XM_017007654.1 | XP_016863143.1 | 3 | 468 | |||
| GenBank mRNA | AY594282.1 | - | 3 | 288 | ||
| KU178526.1 | - | 3 | 288 | |||
| KU178527.1 | - | 3 | 288 | |||

SLC30A9
Genomic Map