siRNA Details
Gene Symbol: Ppp1cc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013636.3 | NP_038664.2 | 4, 5 | 810 | ||
GenBank mRNA | AK076009.1 | - | 4, 5 | 747 | ||
AK088229.1 | - | 4, 5 | 754 | |||
AK147131.1 | - | 4, 5 | 812 | |||
AK152422.1 | - | 4, 5 | 773 | |||
AK152625.1 | - | 4, 5 | 749 | |||
AK153363.1 | - | 4, 5 | 623 | |||
AK169336.1 | - | 4, 5 | 770 | |||
BC008157.1 | - | 4, 5 | 728 | |||
BC010613.1 | - | 4, 5 | 583 | 3589168 | ||
BC021646.1 | - | 4, 5 | 811 | 3487479 | ||
BC039972.1 | - | 4, 5 | 743 | |||
BC085496.1 | - | 4, 5 | 749 | 6829867 | ||
D85137.1 | - | 4, 5 | 635 | |||
M27071.1 | - | 4, 5 | 731 | |||
U53456.1 | - | 4, 5 | 727 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_022498, NM_001270983, XM_008769212, BC078825, FM122849, and others.
Panther Classification:
Gene Ontology Categories:
