siRNA Details
| Gene Symbol: Aff3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001290814.1 | NP_001277743.1 | 18, 19 | 3301 | ||
| GenBank mRNA | BC052061.1 | - | 18, 19 | 2631 | ||
| BQ443842.1 | - | 2, 3 | 141 | |||
| BU056443.1 | - | 5, 6 | 608 | |||

Aff3
Genomic Map