siRNA Details
| Gene Symbol: BMX | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001320866.1 | NP_001307795.1 | 14, 15 | 1562 | ||
| NM_001721.6 | NP_001712.1 | 14, 15 | 1503 | |||
| NM_203281.2 | NP_975010.1 | 14, 15 | 1565 | |||
| GenBank mRNA | AF045459.1 | - | 14, 15 | 1576 | ||
| AK291824.1 | - | 14, 15 | 1535 | |||
| AK313568.1 | - | 14, 15 | 1498 | |||
| BC016652.2 | - | 14, 15 | 1520 | 3919253 | ||
| X83107.1 | - | 14, 15 | 1425 | |||

BMX
Genomic Map