siRNA Details
| Gene Symbol: Gm20767 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039648.3 | NP_001034737.2 | 3 | 954 | ||
| GenBank mRNA | AF067058.1 | - | 2 | 402 | ||
| AK139926.1 | - | 3 | 955 | |||
| BC147576.1 | - | 2 | 434 | 9056566 | ||
| BC147578.1 | - | 2 | 434 | 9056568 | ||

Gm20767
Genomic Map