siRNA Details
| Gene Symbol: Igkv1-135 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | Z95479.1 | - | 535 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Igkv1-135| Gene Symbol: Igkv1-135 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | Z95479.1 | - | 535 | |||