siRNA Details
| Gene Symbol: Ngb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001294308.1 | NP_001281237.1 | 4 | 1476 | ||
| NM_022414.2 | NP_071859.1 | 4 | 1464 | |||
| GenBank mRNA | AK140173.1 | - | 4 | 1460 | ||
| AK165775.1 | - | 4 | 1270 | |||
| BC024263.1 | - | 4 | 1464 | 5133442 | ||

Ngb
Genomic Map