siRNA Details
| Gene Symbol: Lsm7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025349.2 | NP_079625.1 | 4 | 213 | ||
| GenBank mRNA | AK003062.1 | - | 4 | 201 | ||
| AK007462.1 | - | 4 | 210 | |||
| AK009317.1 | - | 4 | 200 | |||
| AK012446.1 | - | 4 | 210 | |||
| AK013355.1 | - | 4 | 214 | |||
| BC027511.1 | - | 4 | 203 | 1382565 | ||
| BC081444.1 | - | 3 | 196 | 5711629 | ||

Lsm7
Genomic Map