siRNA Details
| Gene Symbol: Mrps36 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001190264.1 | NP_001177193.1 | 2 | 152 | ||
| NM_025369.3 | NP_079645.1 | 2 | 152 | |||
| GenBank mRNA | AK003111.1 | - | 2 | 82 | ||
| AK003765.1 | - | 2 | 129 | |||
| AK007512.1 | - | 2 | 123 | |||
| AK009050.1 | - | 2 | 131 | |||
| AK009966.1 | - | 2 | 83 | |||
| BC028499.1 | - | 2 | 108 | 1382215 | ||
| BC096576.1 | - | 2 | 117 | 1401014 | ||

Mrps36
Genomic Map