siRNA Details
| Gene Symbol: Pgls | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001294269.1 | NP_001281198.1 | 2 | 698 | ||
| NM_025396.3 | NP_079672.1 | 2 | 414 | |||
| GenBank mRNA | AK003134.1 | - | 2 | 411 | ||
| AK003985.1 | - | 2 | 415 | |||
| AK028344.1 | - | 1 | 1972 | |||
| AK036059.1 | - | 415 | ||||
| AK172066.1 | - | 2 | 415 | |||
| BC006594.1 | - | 2 | 388 | 3964951 | ||
| BE654173.1 | - | 2 | 276 | |||

Pgls
Genomic Map