siRNA Details
| Gene Symbol: Nat9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025400.3 | NP_079676.1 | 7 | 964 | ||
| GenBank mRNA | AK003948.1 | - | 7 | 918 | ||
| AK009262.1 | - | 6 | 694 | |||
| AK020083.1 | - | 7 | 947 | |||
| AK082469.1 | - | 7 | 904 | |||
| AK148048.1 | - | 7 | 996 | |||
| AK154593.1 | - | 6 | 1095 | |||
| AK213540.1 | - | 1 | 200 | |||
| AK215096.1 | - | 1 | 208 | |||
| BC011315.1 | - | 7 | 964 | 4013700 | ||

Nat9
Genomic Map