siRNA Details
| Gene Symbol: 1110037F02Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081183.1 | NP_001074652.1 | 11 | 2857 | ||
| GenBank mRNA | AK034232.1 | - | 10 | 2629 | ||
| AK052499.1 | - | 10 | 2645 | |||
| AK076731.1 | - | 10 | 2677 | |||
| AK122509.1 | - | 10 | 2610 | |||
| AK155623.1 | - | 10 | 2708 | |||
| AK170075.1 | - | 10 | 2710 | |||
| BC033309.1 | - | 11 | 2827 | 5024976 | ||

1110037F02Rik
Genomic Map