siRNA Details
Gene Symbol: Nenf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025424.2 | NP_079700.1 | 2 | 239 | ||
GenBank mRNA | AB126218.1 | - | 2 | 213 | ||
AK002413.1 | - | 2 | 240 | |||
AK004329.1 | - | 2 | 228 | |||
AK009304.1 | - | 2 | 230 | |||
AK020403.1 | - | 2 | 221 | |||
AK155319.1 | - | 2 | 238 | |||
BC048464.1 | - | 2 | 227 | 6596336 |

More Information
Other Species Targeted

Panther Classification:
Gene Ontology Categories:
