siRNA Details
| Gene Symbol: Aamdc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001177945.1 | NP_001171416.1 | 5 | 775 | ||
| NM_001177946.1 | NP_001171417.1 | 4 | 679 | |||
| NM_001177947.1 | NP_001171418.1 | 3 | 575 | |||
| NM_183251.3 | NP_899074.2 | 4 | 671 | |||
| GenBank mRNA | BC026557.1 | - | 5 | 693 | 5027461 | |
| BC062811.1 | - | 4 | 634 | 1264501 | ||

Aamdc
Genomic Map