siRNA Details
| Gene Symbol: Tmem208 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025486.2 | NP_079762.1 | 6 | 614 | ||
| GenBank mRNA | AK002499.1 | - | 6 | 602 | ||
| AK005662.1 | - | 6 | 598 | |||
| AK218712.1 | - | 1 | 90 | |||
| BC005665.1 | - | 6 | 570 | |||
| BC024608.1 | - | 6 | 565 | 4242513 | ||

Tmem208
Genomic Map