siRNA Details
| Gene Symbol: Fam229b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_183254.1 | NP_899077.1 | 4 | 468 | ||
| GenBank mRNA | AK006353.1 | - | 4 | 468 | ||
| AK006431.1 | - | 4 | 447 | |||
| BC094255.1 | - | 3 | 2217 | |||
| BC147171.1 | - | 3 | 397 | 9056241 | ||
| BC147172.1 | - | 3 | 397 | 9056208 | ||
| BC172004.1 | - | 3 | 394 | 9054290 | ||

Fam229b
Genomic Map