siRNA Details
| Gene Symbol: Cdc26 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_139291.3 | NP_647452.1 | 4 | 605 | ||
| GenBank mRNA | AK004518.1 | - | 1 | 245 | ||
| AK008199.1 | - | 1 | 89 | |||
| AK012627.1 | - | 4 | 606 | |||
| AK047055.1 | - | 625 | ||||
| AK086692.1 | - | 1084 | ||||
| AK135556.1 | - | 605 | ||||
| BC005775.1 | - | 3 | 532 | 3602527 | ||
| BC114350.2 | - | 2 | 392 | 40053418 | ||

Cdc26
Genomic Map