siRNA Details
| Gene Symbol: Exosc7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081188.1 | NP_001074657.1 | 3 | 250 | ||
| GenBank mRNA | AK011294.1 | - | 3 | 237 | ||
| AK129057.1 | - | 3 | 202 | |||
| BC039655.1 | - | 3 | 235 | |||
| BC052656.1 | - | 3 | 232 | 30041585 | ||
| BC094932.1 | - | 3 | 243 | 30008071 | ||
| BC132363.1 | - | 3 | 265 | 40130640 | ||
| BC145675.1 | - | 3 | 265 | 40130978 | ||

Exosc7
Genomic Map